Learn More
ABclonal Technology SRPRB Rabbit pAb
SDP
Supplier:  ABclonal Technology A105911000UL
Encompass_Preferred
The protein encoded by this gene has similarity to mouse protein which is a subunit of the signal recognition particle receptor (SR). This subunit is a transmembrane GTPase belonging to the GTPase superfamily. It anchors alpha subunit, a peripheral membrane GTPase, to the ER membrane. SR is required for the cotranslational targeting of both secretory and membrane proteins to the ER membrane.
Catalog No. 50-001-35780