Learn More
ABclonal Technology ACBD5 Rabbit pAb
SDP
Supplier:  ABclonal Technology A105951000UL
Encompass_Preferred
This gene encodes a member of the acyl-Coenzyme A binding protein family, known to function in the transport and distribution of long chain acyl-Coenzyme A in cells. This gene may play a role in the differentiation of megakaryocytes and formation of platelets. A related protein in yeast is involved in autophagy of peroxisomes. A mutation in this gene has been associated with autosomal dominant thrombocytopenia. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-37885