Learn More
ABclonal Technology Gsdma Rabbit pAb
SDP
Supplier:  ABclonal Technology A106021000UL
Encompass_Preferred
Predicted to enable phosphatidylinositol-4,5-bisphosphate binding activity, phosphatidylinositol-4-phosphate binding activity, and phosphatidylserine binding activity. Predicted to be involved in apoptotic process, defense response to bacterium, and pyroptosis. Predicted to act upstream of or within programmed cell death. Located in cytoplasm. Is expressed in esophagus epithelium, stomach, stomach cardiac region, stomach glandular region, and stomach glandular region mucosa. Orthologous to human GSDMA (gasdermin A).
Catalog No. 50-001-35777