Learn More
ABclonal Technology GCGR Rabbit pAb
SDP
Catalog No. 5000138566
Encompass_Preferred
The protein encoded by this gene is a glucagon receptor that is important in controlling blood glucose levels. Defects in this gene are a cause of non-insulin-dependent diabetes mellitus (NIDDM).
Catalog No. 50-001-38566 Supplier ABclonal Technology Supplier No. A10617500UL
The protein encoded by this gene is a glucagon receptor that is important in controlling blood glucose levels. Defects in this gene are a cause of non-insulin-dependent diabetes mellitus (NIDDM).