Learn More
ABclonal Technology RNF114 Rabbit pAb
SDP
Catalog No. 5000137896
Encompass_Preferred
Predicted to enable ubiquitin protein ligase activity. Predicted to be involved in protein polyubiquitination and ubiquitin-dependent protein catabolic process. Located in cytosol and plasma membrane. Biomarker of male infertility.
Catalog No. 50-001-37896 Supplier ABclonal Technology Supplier No. A10636500UL
Predicted to enable ubiquitin protein ligase activity. Predicted to be involved in protein polyubiquitination and ubiquitin-dependent protein catabolic process. Located in cytosol and plasma membrane. Biomarker of male infertility.