Learn More
ABclonal Technology TMPRSS6 Rabbit pAb
SDP
Catalog No. 5000138617
Encompass_Preferred
The protein encoded by this gene is a type II transmembrane serine proteinase that is found attached to the cell surface. The encoded protein may be involved in matrix remodeling processes in the liver. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-38617 Supplier ABclonal Technology Supplier No. A106421000UL
The protein encoded by this gene is a type II transmembrane serine proteinase that is found attached to the cell surface. The encoded protein may be involved in matrix remodeling processes in the liver. Alternative splicing results in multiple transcript variants.