Learn More
ABclonal Technology CALCR Rabbit pAb
SDP
Supplier:  ABclonal Technology A10674100UL
Encompass_Preferred
This gene encodes a high affinity receptor for the peptide hormone calcitonin and belongs to a subfamily of seven transmembrane-spanning G protein-coupled receptors. The encoded protein is involved in maintaining calcium homeostasis and in regulating osteoclast-mediated bone resorption. Polymorphisms in this gene have been associated with variations in bone mineral density and onset of osteoporosis. Alternate splicing results in multiple transcript variants.
Catalog No. 50-001-37840