Learn More
ABclonal Technology CYBA Rabbit pAb
SDP
Supplier:  ABclonal Technology A106941000UL
Encompass_Preferred
Cytochrome b is comprised of a light chain (alpha) and a heavy chain (beta). This gene encodes the light, alpha subunit which has been proposed as a primary component of the microbicidal oxidase system of phagocytes. Mutations in this gene are associated with autosomal recessive chronic granulomatous disease (CGD), that is characterized by the failure of activated phagocytes to generate superoxide, which is important for the microbicidal activity of these cells.
Catalog No. 50-001-37737