Learn More
ABclonal Technology DEFB132 Rabbit pAb
SDP
Catalog No. 5000137788
Encompass_Preferred
Defensins are cysteine-rich cationic polypeptides that are important in the immunologic response to invading microorganisms. The protein encoded by this gene is secreted and is a member of the beta defensin protein family. This protein binds spermatozoa and has antimicrobial activity against E. coli. Beta defensin genes are found in several clusters throughout the genome, with this gene mapping to a cluster at 20p13.
Catalog No. 50-001-37788 Supplier ABclonal Technology Supplier No. A1074500UL
Defensins are cysteine-rich cationic polypeptides that are important in the immunologic response to invading microorganisms. The protein encoded by this gene is secreted and is a member of the beta defensin protein family. This protein binds spermatozoa and has antimicrobial activity against E. coli. Beta defensin genes are found in several clusters throughout the genome, with this gene mapping to a cluster at 20p13.