Learn More
ABclonal Technology LAPTM4B Rabbit pAb
SDP
Catalog No. 5000138254
Encompass_Preferred
Enables ceramide binding activity, enzyme binding activity, and phosphatidylinositol bisphosphate binding activity. Involved in several processes, including negative regulation of macromolecule metabolic process, regulation of lysosomal membrane permeability, and regulation of lysosome organization. Located in several cellular components, including endosome, lysosomal membrane, and plasma membrane.
Catalog No. 50-001-38254 Supplier ABclonal Technology Supplier No. A10761500UL
Enables ceramide binding activity, enzyme binding activity, and phosphatidylinositol bisphosphate binding activity. Involved in several processes, including negative regulation of macromolecule metabolic process, regulation of lysosomal membrane permeability, and regulation of lysosome organization. Located in several cellular components, including endosome, lysosomal membrane, and plasma membrane.