Learn More
ABclonal Technology ZP1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10776500UL
Encompass_Preferred
The zona pellucida is an extracellular matrix that surrounds the oocyte and early embryo. It is composed primarily of three or four glycoproteins with various functions during fertilization and preimplantation development. The protein encoded by this gene ensures the structural integrity of the zona pellucida. Mutations in this gene are a cause of oocyte maturation defect and infertility.
Catalog No. 50-001-37812