Learn More
ABclonal Technology RBM5 Rabbit pAb
SDP
Supplier:  ABclonal Technology A107881000UL
Encompass_Preferred
This gene is a candidate tumor suppressor gene which encodes a nuclear RNA binding protein that is a component of the spliceosome A complex. The encoded protein plays a role in the induction of cell cycle arrest and apoptosis through pre-mRNA splicing of multiple target genes including the tumor suppressor protein p53. This gene is located within the tumor suppressor region 3p21.3, and may play a role in the inhibition of tumor transformation and progression of several malignancies including lung cancer.
Catalog No. 50-001-37373