Learn More
ABclonal Technology MELK Rabbit pAb
SDP
Supplier:  ABclonal Technology A107941000UL
Encompass_Preferred
Enables calcium ion binding activity, non-membrane spanning protein tyrosine kinase activity, and protein serine/threonine kinase activity. Involved in apoptotic process, cell population proliferation, and protein autophosphorylation. Located in cell cortex and plasma membrane.
Catalog No. 50-001-35747