Learn More
ABclonal Technology UCK2 Rabbit pAb
SDP
Catalog No. 5000137743
Encompass_Preferred
This gene encodes a pyrimidine ribonucleoside kinase. The encoded protein (EC 2.7.1.48) catalyzes phosphorylation of uridine and cytidine to uridine monophosphate (UMP) and cytidine monophosphate (CMP), respectively.
Catalog No. 50-001-37743 Supplier ABclonal Technology Supplier No. A108151000UL
This gene encodes a pyrimidine ribonucleoside kinase. The encoded protein (EC 2.7.1.48) catalyzes phosphorylation of uridine and cytidine to uridine monophosphate (UMP) and cytidine monophosphate (CMP), respectively.