Learn More
ABclonal Technology Ambra1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1083500UL
Encompass_Preferred
Enables GTPase binding activity and ubiquitin protein ligase binding activity. Involved in macroautophagy, positive regulation of phosphatidylinositol 3-kinase activity, and response to mitochondrial depolarisation. Located in cytosol. Colocalizes with mitochondrion. Biomarker of multiple system atrophy.
Catalog No. 50-001-37371