Learn More
ABclonal Technology [KO Validated] Hsp90β Rabbit pAb
SDP
Catalog No. 5000136472
Encompass_Preferred
This gene encodes a member of the heat shock protein 90 family, these proteins are involved in signal transduction, protein folding and degradation and morphological evolution. This gene encodes the constitutive form of the cytosolic 90 kDa heat-shock protein and is thought to play a role in gastric apoptosis and inflammation. Alternative splicing results in multiple transcript variants. Pseudogenes have been identified on multiple chromosomes.
Catalog No. 50-001-36472 Supplier ABclonal Technology Supplier No. A1087500UL
This gene encodes a member of the heat shock protein 90 family, these proteins are involved in signal transduction, protein folding and degradation and morphological evolution. This gene encodes the constitutive form of the cytosolic 90 kDa heat-shock protein and is thought to play a role in gastric apoptosis and inflammation. Alternative splicing results in multiple transcript variants. Pseudogenes have been identified on multiple chromosomes.