Learn More
Medchemexpress LLC A11 (acetate) | 2412926-30-6 | 98.5% | 2767.16 | 5 MG
SDP
Catalog No. 5000423695
Encompass_Preferred
A11 acetate is an ANXA1-EphA2 interaction blocker peptide that inhibits nasopharyngeal carcinoma (NPC) cell proliferation, migration, invasion, and angiogenesis.
  • Decreases ANXA1 bound to EphA2
  • Increases Cbl bound to EphA2
  • Decreases EphA2 levels and increases EphA2 ubiquitination
  • Increases EphA2 internalization and colocalization with Cbl in NPC cells
  • Inhibits NPC cell proliferation
  • Inhibits migration, invasion, and angiogenesis
Catalog No. 50-004-23695 Supplier Medchemexpress LLC Supplier No. HYP10412A5MG
A11 acetate is an ANXA1-EphA2 interaction blocker peptide that inhibits nasopharyngeal carcinoma (NPC) cell proliferation, migration, invasion, and angiogenesis.
  • Decreases ANXA1 bound to EphA2
  • Increases Cbl bound to EphA2
  • Decreases EphA2 levels and increases EphA2 ubiquitination
  • Increases EphA2 internalization and colocalization with Cbl in NPC cells
  • Inhibits NPC cell proliferation
  • Inhibits migration, invasion, and angiogenesis