Learn More
Medchemexpress LLC A11 (acetate) | 98.5% | 2767.16 | 25 MG
SDP
Catalog No. 5000426801
Encompass_Preferred
A11 acetate is a chemical compound primarily intended for laboratory use and the manufacturing of substances. Presented as a white to off-white solid, it boasts a high purity of 98.5% and is not classified as a hazardous substance.
  • Suitable for laboratory research and substance manufacturing
  • Not classified as a hazardous substance or mixture
  • High purity of 98.5%
  • Recommended storage: Powder at -80°C for up to 2 years or -20°C for 1 year; in solvent at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)
  • White to off-white solid appearance
Catalog No. 50-004-26801 Supplier Medchemexpress LLC Supplier No. 25MGHYP10412A
A11 acetate is a chemical compound primarily intended for laboratory use and the manufacturing of substances. Presented as a white to off-white solid, it boasts a high purity of 98.5% and is not classified as a hazardous substance.
  • Suitable for laboratory research and substance manufacturing
  • Not classified as a hazardous substance or mixture
  • High purity of 98.5%
  • Recommended storage: Powder at -80°C for up to 2 years or -20°C for 1 year; in solvent at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)
  • White to off-white solid appearance