Learn More
Medchemexpress LLC A11 (acetate) | 98.5% | 2767.16 (free base) | 10 MG
SDP
Catalog No. 5000424707
Encompass_Preferred
A11 acetate is an ANXA1-derived 11 amino acid-long peptide that functions as an ANXA1-EphA2 interaction blocker peptide. It has a molecular weight of 2767.16 (free base) and a purity of 98.5%. This peptide is suitable for laboratory research applications.
  • Functions as an ANXA1-EphA2 interaction blocker peptide
  • 11 amino acid-long peptide
  • High purity (98.5%)
  • Recommended storage conditions for powder and in solvent
Catalog No. 50-004-24707 Supplier Medchemexpress LLC Supplier No. HYP10412A10MG
A11 acetate is an ANXA1-derived 11 amino acid-long peptide that functions as an ANXA1-EphA2 interaction blocker peptide. It has a molecular weight of 2767.16 (free base) and a purity of 98.5%. This peptide is suitable for laboratory research applications.
  • Functions as an ANXA1-EphA2 interaction blocker peptide
  • 11 amino acid-long peptide
  • High purity (98.5%)
  • Recommended storage conditions for powder and in solvent