Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ AH-7921 Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco A1131ML

Catalog No. 01-001-006


Description

Description

This certified Snap-N-Spike™ Solution is suitable for use as starting material in calibrators and controls for AH-7921 LC-MS or GC/MS methods in clinical toxicology, urine drug testing, or forensic analysis applications. AH-7921 is an opioid designer drug with 80% the potency of morphine when administered orally. First discovered in 2013, AH-7921 is an active ingredient of synthetic cannabis found in Japan.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 41804-96-0
MFCD27939621
1.0 mg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
C16H22Cl2N2O · HCl
UN 1230 - class 3 - PG 2 - Methanol
365.73
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation