Learn More
ABclonal Technology HNF4G Rabbit pAb
SDP
Supplier:  ABclonal Technology A160511000UL
Encompass_Preferred
Enables DNA-binding transcription activator activity, RNA polymerase II-specific and RNA polymerase II cis-regulatory region sequence-specific DNA binding activity. Involved in positive regulation of transcription by RNA polymerase II. Located in several cellular components, including intercellular bridge, mitotic spindle, and nucleoplasm.
Catalog No. 50-001-11700