Learn More
ABclonal Technology IGHMBP2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A160531000UL
Encompass_Preferred
This gene encodes a helicase superfamily member that binds a specific DNA sequence from the immunoglobulin mu chain switch region. Mutations in this gene lead to spinal muscle atrophy with respiratory distress type 1.
Catalog No. 50-001-12151