Learn More
ABclonal Technology CCDC6 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16075500UL
Encompass_Preferred
This gene encodes a coiled-coil domain-containing protein. The encoded protein is ubiquitously expressed and may function as a tumor suppressor. A chromosomal rearrangement resulting in the expression of a fusion gene containing a portion of this gene and the intracellular kinase-encoding domain of the ret proto-oncogene is the cause of thyroid papillary carcinoma.
Catalog No. 50-001-10244