Learn More
ABclonal Technology EPN2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16106500UL
Encompass_Preferred
This gene encodes a protein which interacts with clathrin and adaptor-related protein complex 2, alpha 1 subunit. The protein is found in a brain-derived clathrin-coated vesicle fraction and localizes to the peri-Golgi region and the cell periphery. The protein is thought to be involved in clathrin-mediated endocytosis. Alternate splicing of this gene results in multiple transcript variants encoding different isoforms.
Catalog No. 50-001-10243