Learn More
ABclonal Technology PDE11A Rabbit pAb
SDP
Supplier:  ABclonal Technology A161211000UL
Encompass_Preferred
The 3,5-cyclic nucleotides cAMP and cGMP function as second messengers in a wide variety of signal transduction pathways. 3,5-cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and cGMP to the corresponding 5-monophosphates and provide a mechanism to downregulate cAMP and cGMP signaling. This gene encodes a member of the PDE protein superfamily. Mutations in this gene are a cause of Cushing disease and adrenocortical hyperplasia. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-11127