Learn More
ABclonal Technology BPIFB2 Rabbit pAb
SDP
Catalog No. 5000111112
Encompass_Preferred
This gene encodes a member of the lipid transfer/lipopolysaccharide binding protein (LT/LBP) gene family. It is highly expressed in hypertrophic tonsils. This gene and three other members of the LT/LBP gene family form a cluster on the long arm of chromosome 20.
Catalog No. 50-001-11112 Supplier ABclonal Technology Supplier No. A16148500UL
This gene encodes a member of the lipid transfer/lipopolysaccharide binding protein (LT/LBP) gene family. It is highly expressed in hypertrophic tonsils. This gene and three other members of the LT/LBP gene family form a cluster on the long arm of chromosome 20.