Learn More
ABclonal Technology HPS4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A161571000UL
Encompass_Preferred
This gene encodes a protein component of biogenesis of lysosome-related organelles complexes (BLOC). BLOC complexes are important for the formation of endosomal-lysosomal organelles such as melanosomes and platelet dense granules. Mutations in this gene result in subtype 4 of Hermansky-Pudlak syndrome, a form of albinism. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-11667