Learn More
ABclonal Technology ZNF677 Rabbit pAb
SDP
Supplier:  ABclonal Technology A161831000UL
Encompass_Preferred
Predicted to enable DNA-binding transcription factor activity, RNA polymerase II-specific and RNA polymerase II cis-regulatory region sequence-specific DNA binding activity. Predicted to be involved in regulation of transcription by RNA polymerase II. Predicted to be active in nucleus.
Catalog No. 50-001-10479