Learn More
ABclonal Technology USP33 Rabbit pAb
SDP
Catalog No. 5000112596
Encompass_Preferred
This gene encodes a deubiquinating enzyme important in a variety of processes, including Slit-dependent cell migration and beta-2 adrenergic receptor signaling. The protein is negatively regulated through ubiquitination by von Hippel-Lindau tumor protein (VHL). Alternative splicing results in multiple transcript variants and protein isoforms.
Catalog No. 50-001-12596 Supplier ABclonal Technology Supplier No. A161871000UL
Add to Cart
Add to Cart
This gene encodes a deubiquinating enzyme important in a variety of processes, including Slit-dependent cell migration and beta-2 adrenergic receptor signaling. The protein is negatively regulated through ubiquitination by von Hippel-Lindau tumor protein (VHL). Alternative splicing results in multiple transcript variants and protein isoforms.