Learn More
ABclonal Technology MAGOHB Rabbit pAb
SDP
Supplier:  ABclonal Technology A161921000UL
Encompass_Preferred
Enables RNA binding activity. Involved in mRNA splicing, via spliceosome and nuclear-transcribed mRNA catabolic process, nonsense-mediated decay. Located in nucleus. Part of U2-type catalytic step 1 spliceosome, U2-type precatalytic spliceosome, and exon-exon junction complex.
Catalog No. 50-001-11379