Learn More
ABclonal Technology NDRG4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A161951000UL
Encompass_Preferred
This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that is required for cell cycle progression and survival in primary astrocytes and may be involved in the regulation of mitogenic signalling in vascular smooth muscles cells. Alternative splicing results in multiple transcripts encoding different isoforms.
Catalog No. 50-001-10476