Learn More
ABclonal Technology KIAA1429 Rabbit pAb
SDP
Catalog No. 5000111402
Encompass_Preferred
Enables RNA binding activity. Involved in mRNA alternative polyadenylation and mRNA methylation. Located in cytosol and nuclear speck. Colocalizes with RNA N6-methyladenosine methyltransferase complex.
Catalog No. 50-001-11402 Supplier ABclonal Technology Supplier No. A162011000UL
Enables RNA binding activity. Involved in mRNA alternative polyadenylation and mRNA methylation. Located in cytosol and nuclear speck. Colocalizes with RNA N6-methyladenosine methyltransferase complex.