Learn More
ABclonal Technology SLC6A9 Rabbit pAb
SDP
Catalog No. 5000110518
Encompass_Preferred
The amino acid glycine acts as an inhibitory neurotransmitter in the central nervous system. The protein encoded by this gene is one of two transporters that stop glycine signaling by removing it from the synaptic cleft.
Catalog No. 50-001-10518 Supplier ABclonal Technology Supplier No. A16203500UL
The amino acid glycine acts as an inhibitory neurotransmitter in the central nervous system. The protein encoded by this gene is one of two transporters that stop glycine signaling by removing it from the synaptic cleft.