Learn More
ABclonal Technology SDHAF2 Rabbit pAb
SDP
Catalog No. 5000110660
Encompass_Preferred
This gene encodes a mitochondrial assembly factor needed for the flavination of a succinate dehydrogenase complex subunit (SDHA), which is required for activity of the succinate dehydrogenase complex. Mutations in this gene are associated with paraganglioma.
Catalog No. 50-001-10660 Supplier ABclonal Technology Supplier No. A16204500UL
This gene encodes a mitochondrial assembly factor needed for the flavination of a succinate dehydrogenase complex subunit (SDHA), which is required for activity of the succinate dehydrogenase complex. Mutations in this gene are associated with paraganglioma.