Learn More
ABclonal Technology FABPI Rabbit pAb
SDP
Supplier:  ABclonal Technology A16211000UL
Encompass_Preferred
The protein encoded by this gene is an intracellular fatty acid-binding protein that participates in the uptake, intracellular metabolism, and transport of long-chain fatty acids. The encoded protein is also involved in the modulation of cell growth and proliferation. This protein binds saturated long-chain fatty acids with high affinity, and may act as a lipid sensor to maintain energy homeostasis.
Catalog No. 50-001-12590