Learn More
ABclonal Technology Casein Kinase 1 alpha (CSNK1A1) Rabbit pAb
SDP
Catalog No. 5000112587
Encompass_Preferred
Enables protein serine/threonine kinase activity. Involved in several processes, including negative regulation of canonical Wnt signaling pathway, peptidyl-serine phosphorylation, and positive regulation of proteasomal ubiquitin-dependent protein catabolic process. Located in centrosome, cytosol, and nuclear speck. Part of beta-catenin destruction complex. Colocalizes with keratin filament and mRNA cleavage and polyadenylation specificity factor complex. Biomarker of Alzheimers disease and inclusion body myositis.
Catalog No. 50-001-12587 Supplier ABclonal Technology Supplier No. A162251000UL
Enables protein serine/threonine kinase activity. Involved in several processes, including negative regulation of canonical Wnt signaling pathway, peptidyl-serine phosphorylation, and positive regulation of proteasomal ubiquitin-dependent protein catabolic process. Located in centrosome, cytosol, and nuclear speck. Part of beta-catenin destruction complex. Colocalizes with keratin filament and mRNA cleavage and polyadenylation specificity factor complex. Biomarker of Alzheimers disease and inclusion body myositis.