Learn More
ABclonal Technology ID4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A162291000UL
Encompass_Preferred
This gene encodes a member of the inhibitor of DNA binding (ID) protein family. The encoded protein lacks DNA binding ability, and instead regulates gene expression through binding to and inhibiting basic helix-loop-helix transcription factors. This protein has been implicated in the regulation of diverse cellular processes that play a role in development and tumorigenesis.
Catalog No. 50-001-10610