Learn More
ABclonal Technology MRPL18 Rabbit pAb
SDP
Supplier:  ABclonal Technology A162311000UL
Encompass_Preferred
This nuclear gene encodes a protein component of the larger 39S subunit of mitochondrial ribosome. This protein may also aid in the import of nuclear-encoded 5S rRNA into mitochondria. Alternative splicing results in multiple transcript variants, most of which are not predicted to encode a protein. A pseudogene of this gene is found on chromosome 16.
Catalog No. 50-001-10611