Learn More
ABclonal Technology GGPS1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16233500UL
Encompass_Preferred
This gene is a member of the prenyltransferase family and encodes a protein with geranylgeranyl diphosphate (GGPP) synthase activity. The enzyme catalyzes the synthesis of GGPP from farnesyl diphosphate and isopentenyl diphosphate. GGPP is an important molecule responsible for the C20-prenylation of proteins and for the regulation of a nuclear hormone receptor. Alternate transcriptional splice variants, both protein-coding and non-protein-coding, have been found for this gene.
Catalog No. 50-001-10449