Learn More
ABclonal Technology FOXL2 Rabbit pAb
SDP
Catalog No. 5000113423
Encompass_Preferred
This gene encodes a forkhead transcription factor. The protein contains a fork-head DNA-binding domain and may play a role in ovarian development and function. Expansion of a polyalanine repeat region and other mutations in this gene are a cause of blepharophimosis syndrome and premature ovarian failure 3.
Catalog No. 50-001-13423 Supplier ABclonal Technology Supplier No. A16244500UL
This gene encodes a forkhead transcription factor. The protein contains a fork-head DNA-binding domain and may play a role in ovarian development and function. Expansion of a polyalanine repeat region and other mutations in this gene are a cause of blepharophimosis syndrome and premature ovarian failure 3.