Learn More
ABclonal Technology SMPD1 / ASM Rabbit pAb
SDP
Supplier:  ABclonal Technology A16263500UL
Encompass_Preferred
The protein encoded by this gene is a lysosomal acid sphingomyelinase that converts sphingomyelin to ceramide. The encoded protein also has phospholipase C activity. Defects in this gene are a cause of Niemann-Pick disease type A (NPA) and Niemann-Pick disease type B (NPB). Multiple transcript variants encoding different isoforms have been identified.
Catalog No. 50-001-10810