Learn More
ABclonal Technology DDX6 Rabbit pAb
SDP
Supplier:  ABclonal Technology A162701000UL
Encompass_Preferred
This gene encodes a member of the DEAD box protein family. The protein is an RNA helicase found in P-bodies and stress granules, and functions in translation suppression and mRNA degradation. It is required for microRNA-induced gene silencing. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Catalog No. 50-001-09917