Learn More
ABclonal Technology FUT4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A163201000UL
Encompass_Preferred
The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15).
Catalog No. 50-001-10434