Learn More
ABclonal Technology FUT4 Rabbit pAb
SDP
Catalog No. 5000109867
Encompass_Preferred
The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15).
Catalog No. 50-001-09867 Supplier ABclonal Technology Supplier No. A16320500UL
The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15).