Learn More
ABclonal Technology DUSP15 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16322500UL
Encompass_Preferred
The protein encoded by this gene has both protein-tyrosine phophatase activity and serine/threonine-specific phosphatase activity, and therefore is known as a dual specificity phosphatase. This protein may function in the differentiation of oligodendrocytes. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-10839