Learn More
ABclonal Technology AEBP1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A163401000UL
Encompass_Preferred
This gene encodes a member of carboxypeptidase A protein family. The encoded protein may function as a transcriptional repressor and play a role in adipogenesis and smooth muscle cell differentiation. Studies in mice suggest that this gene functions in wound healing and abdominal wall development. Overexpression of this gene is associated with glioblastoma.
Catalog No. 50-001-11936