Learn More
ABclonal Technology ETV6 Rabbit pAb
SDP
Supplier:  ABclonal Technology A16371000UL
Encompass_Preferred
This gene encodes an ETS family transcription factor. The product of this gene contains two functional domains a N-terminal pointed (PNT) domain that is involved in protein-protein interactions with itself and other proteins, and a C-terminal DNA-binding domain. Gene knockout studies in mice suggest that it is required for hematopoiesis and maintenance of the developing vascular network. This gene is known to be involved in a large number of chromosomal rearrangements associated with leukemia and congenital fibrosarcoma.
Catalog No. 50-001-10871