Learn More
ABclonal Technology ENO3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A163711000UL
Encompass_Preferred
This gene encodes one of the three enolase isoenzymes found in mammals. This isoenzyme is found in skeletal muscle cells in the adult where it may play a role in muscle development and regeneration. A switch from alpha enolase to beta enolase occurs in muscle tissue during development in rodents. Mutations in this gene have be associated glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been described.
Catalog No. 50-001-10512