Learn More
ABclonal Technology FGD1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A163741000UL
Encompass_Preferred
This gene encodes a protein that contains Dbl (DH) and pleckstrin (PH) homology domains and is similar to the Rho family of small GTP-binding proteins. The encoded protein specifically binds to the Rho family GTPase Cdc42Hs and can stimulate the GDP-GTP exchange of the isoprenylated form of Cdc42Hs. It also stimulates the mitogen activated protein kinase cascade leading to c-Jun kinase SAPK/JNK1 activation. Defects in this gene are the cause of the faciogenital dysplasia in Aarskog-Scott syndrome and a syndromatic form of X-linked cognitive disability.
Catalog No. 50-001-12051