Learn More
ABclonal Technology FKBP1B Rabbit pAb
SDP
Supplier:  ABclonal Technology A163751000UL
Encompass_Preferred
The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin. It is highly similar to the FK506-binding protein 1A. Its physiological role is thought to be in excitation-contraction coupling in cardiac muscle. There are two alternatively spliced transcript variants of this gene encoding different isoforms.
Catalog No. 50-001-10485